Assay Details
Gene Symbol: Rgs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019341.1 | NP_062214.1 | 4 - 5 | 391 | 103 | |
GenBank mRNA | AF241259.1 | - | 4 - 5 | 391 | 103 | |
EF568934.1 | - | 4 - 5 | 455 | 103 | ||
U32435.1 | - | 1 - 2 | 109 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
