Assay Details
Gene Symbol: Cxcl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022214.1 | NP_071550.1 | 3 - 4 | 331 | 73 | |
XM_008770055.1 | XP_008768277.1 | 10 - 11 | 1725 | 73 | ||
GenBank mRNA | U90448.1 | - | 3 - 4 | 331 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
