Assay Details
Gene Symbol: Clpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022947.1 | NP_075236.1 | 10 - 11 | 1218 | 69 | |
XM_008759796.1 | XP_008758018.1 | 11 - 12 | 1572 | 69 | ||
GenBank mRNA | AB027570.1 | - | 10 - 11 | 1218 | 69 | |
BC070917.1 | - | 10 - 11 | 1181 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
