Assay Details
Gene Symbol: Clpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022947.1 | NP_075236.1 | 13 - 14 | 1602 | 103 | |
XM_008759796.1 | XP_008758018.1 | 14 - 15 | 1956 | 103 | ||
GenBank mRNA | AB027570.1 | - | 13 - 14 | 1602 | 103 | |
BC070917.1 | - | 13 - 14 | 1565 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
