Assay Details
Gene Symbol: Clpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022947.1 | NP_075236.1 | 3 - 4 | 588 | 76 | |
XM_008759796.1 | XP_008758018.1 | 3 - 4 | 852 | 76 | ||
GenBank mRNA | AB027570.1 | - | 3 - 4 | 588 | 76 | |
BC070917.1 | - | 3 - 4 | 551 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
