Assay Details
Gene Symbol: Hap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024133.2 | NP_077047.1 | 2 - 3 | 499 | 86 | |
NM_177982.1 | NP_817091.1 | 2 - 3 | 499 | 86 | ||
GenBank mRNA | U38370.1 | - | 2 - 3 | 514 | 86 | |
U38373.1 | - | 2 - 3 | 514 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
