Assay Details
Gene Symbol: Gch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024356.1 | NP_077332.1 | 5 - 6 | 721 | 88 | |
GenBank mRNA | FQ220953.1 | - | 5 - 6 | 708 | 88 | |
M58364.1 | - | 5 - 6 | 721 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
