Assay Details
Gene Symbol: Pdgfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031317.1 | NP_112607.1 | 3 - 4 | 498 | 77 | |
GenBank mRNA | AB033830.1 | - | 3 - 4 | 498 | 77 | |
AF508348.1 | - | 2 - 3 | 376 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
