Assay Details
Gene Symbol: Tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031326.1 | NP_112616.1 | 3 - 4 | 394 | 78 | |
GenBank mRNA | AB014089.1 | - | 3 - 4 | 394 | 78 | |
AF246196.1 | - | 2 - 3 | 171 | 78 | ||
AF377866.1 | - | 3 - 4 | 401 | 78 | ||
AJ312746.1 | - | 3 - 4 | 394 | 78 | ||
BC062022.1 | - | 3 - 4 | 396 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
