Assay Details
Gene Symbol: Tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031326.1 | NP_112616.1 | 6 - 7 | 698 | 96 | |
GenBank mRNA | AB014089.1 | - | 6 - 7 | 698 | 96 | |
AF246196.1 | - | 5 - 6 | 475 | 96 | ||
AF377866.1 | - | 6 - 7 | 705 | 96 | ||
AJ312746.1 | - | 6 - 7 | 698 | 96 | ||
BC062022.1 | - | 6 - 7 | 700 | 96 | ||
FQ219683.1 | - | 3 - 4 | 255 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
