Assay Details
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031332.1 | NP_112622.1 | 2 - 3 | 458 | 69 | |
GenBank mRNA | AB017446.1 | - | 2 - 3 | 458 | 69 | |
BC081777.1 | - | 2 - 3 | 456 | 69 | ||
FQ209755.1 | - | 470 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
