Assay Details
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031332.1 | NP_112622.1 | 4 - 5 | 719 | 93 | |
XM_008760243.1 | XP_008758465.1 | 3 - 4 | 534 | 93 | ||
GenBank mRNA | AB017446.1 | - | 4 - 5 | 719 | 93 | |
BC081777.1 | - | 4 - 5 | 717 | 93 | ||
FQ209755.1 | - | 731 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
