Assay Details
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031332.1 | NP_112622.1 | 7 - 8 | 1127 | 63 | |
XM_008760243.1 | XP_008758465.1 | 6 - 7 | 942 | 63 | ||
GenBank mRNA | AB017446.1 | - | 7 - 8 | 1127 | 63 | |
BC081777.1 | - | 7 - 8 | 1125 | 63 | ||
FQ209755.1 | - | 1139 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
