Assay Details
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031354.1 | NP_112644.1 | 7 - 8 | 910 | 107 | |
XM_006251661.1 | XP_006251723.1 | 8 - 9 | 1031 | 107 | ||
GenBank mRNA | AB039663.1 | - | 7 - 8 | 910 | 107 | |
AF268468.1 | - | 6 - 7 | 746 | 107 | ||
BC063164.1 | - | 7 - 8 | 879 | 107 | ||
FQ214165.1 | - | 4 - 5 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
