Assay Details
Gene Symbol: Csnk1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031617.1 | NP_113805.1 | 1 - 2 | 82 | 67 | |
XM_006242038.2 | XP_006242100.1 | 2 - 3 | 231 | 67 | ||
GenBank mRNA | AB042191.1 | - | 1 - 2 | 82 | 67 | |
AB042192.1 | - | 1 - 2 | 82 | 67 | ||
AB056113.1 | - | 1 - 2 | 82 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
