Assay Details
Gene Symbol: Csnk1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031617.1 | NP_113805.1 | 2 - 3 | 187 | 69 | |
XM_006242038.2 | XP_006242100.1 | 3 - 4 | 336 | 69 | ||
GenBank mRNA | AB042191.1 | - | 2 - 3 | 187 | 69 | |
AB042192.1 | - | 2 - 3 | 187 | 69 | ||
AB056113.1 | - | 2 - 3 | 187 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
