Assay Details
Gene Symbol: Csnk1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031617.1 | NP_113805.1 | 7 - 8 | 1080 | 60 | |
XM_006242038.2 | XP_006242100.1 | 8 - 9 | 1229 | 60 | ||
GenBank mRNA | AB042191.1 | - | 7 - 8 | 1080 | 60 | |
AB056113.1 | - | 7 - 8 | 1080 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
