Assay Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031665.2 | NP_113853.1 | 2 - 3 | 371 | 96 | |
GenBank mRNA | BC081769.1 | - | 2 - 3 | 371 | 96 | |
U56815.1 | - | 2 - 3 | 347 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
