Assay Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031665.2 | NP_113853.1 | 5 - 6 | 656 | 87 | |
GenBank mRNA | BC081769.1 | - | 5 - 6 | 656 | 87 | |
U56815.1 | - | 5 - 6 | 632 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
