Assay Details
Gene Symbol: Atp6ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031785.1 | NP_113973.1 | 5 - 6 | 588 | 84 | |
GenBank mRNA | AF035387.1 | - | 5 - 6 | 588 | 84 | |
BC070937.1 | - | 5 - 6 | 588 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
