Assay Details
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031816.1 | NP_114004.1 | 10 - 11 | 1368 | 69 | |
GenBank mRNA | AF090306.1 | - | 10 - 11 | 1368 | 69 | |
BC062012.1 | - | 10 - 11 | 1277 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
