Assay Details
Gene Symbol: Pnliprp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032081.1 | NP_114470.1 | 7 - 8 | 761 | 61 | |
GenBank mRNA | X61925.1 | - | 7 - 8 | 761 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
