Assay Details
Gene Symbol: Dffb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053362.1 | NP_445814.1 | 1 - 2 | 136 | 76 | |
GenBank mRNA | AF136598.1 | - | 1 - 2 | 136 | 76 | |
BC129091.1 | - | 165 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
