Assay Details
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053427.1 | NP_445879.1 | 9 - 10 | 1643 | 79 | |
XM_006229265.1 | XP_006229327.1 | 9 - 10 | 1172 | 79 | ||
GenBank mRNA | AF271235.1 | - | 9 - 10 | 1643 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
