Assay Details
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053486.1 | NP_445938.1 | 4 - 5 | 1969 | 108 | |
GenBank mRNA | AF083330.1 | - | 4 - 5 | 1939 | 108 | |
AJ223599.1 | - | 4 - 5 | 1969 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
