Assay Details
Gene Symbol: Magt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053946.1 | NP_446398.1 | 4 - 5 | 584 | 76 | |
GenBank mRNA | AF008554.2 | - | 4 - 5 | 584 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
