Assay Details
Gene Symbol: Ptpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053990.1 | NP_446442.1 | 2 - 3 | 175 | 99 | |
XM_006254854.2 | XP_006254916.1 | 2 - 3 | 1414 | 99 | ||
XM_006254855.2 | XP_006254917.1 | 2 - 3 | 1414 | 99 | ||
XM_008772080.1 | XP_008770302.1 | 2 - 3 | 1414 | 99 | ||
XM_008772081.1 | XP_008770303.1 | 2 - 3 | 1414 | 99 | ||
GenBank mRNA | BC078758.1 | - | 2 - 3 | 214 | 99 | |
X58828.1 | - | 2 - 3 | 175 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
