Assay Details
Gene Symbol: Ptpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053990.1 | NP_446442.1 | 8 - 9 | 985 | 82 | |
XM_006254855.2 | XP_006254917.1 | 8 - 9 | 2281 | 82 | ||
XM_008772081.1 | XP_008770303.1 | 8 - 9 | 2281 | 82 | ||
GenBank mRNA | AF502573.1 | - | 1 - 2 | 143 | 82 | |
BC078758.1 | - | 8 - 9 | 1024 | 82 | ||
X58828.1 | - | 8 - 9 | 985 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
