Assay Details
Gene Symbol: Scarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054001.2 | NP_446453.1 | 10 - 11 | 1528 | 82 | |
GenBank mRNA | BC061853.1 | - | 10 - 11 | 1528 | 82 | |
D10587.1 | - | 10 - 11 | 1467 | 82 | ||
M68965.1 | - | 10 - 11 | 1476 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
