Assay Details
Gene Symbol: Scarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054001.2 | NP_446453.1 | 7 - 8 | 1281 | 74 | |
GenBank mRNA | BC061853.1 | - | 7 - 8 | 1281 | 74 | |
D10587.1 | - | 7 - 8 | 1220 | 74 | ||
M68965.1 | - | 7 - 8 | 1229 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
