Assay Details
Gene Symbol: Ppp2r2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057116.1 | NP_476457.1 | 333 | 83 | ||
GenBank mRNA | D38261.1 | - | 333 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
