Assay Details
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080896.1 | NP_543172.1 | 10 - 11 | 1133 | 107 | |
GenBank mRNA | AF367468.1 | - | 10 - 11 | 1133 | 107 | |
BC099792.1 | - | 1182 | 107 | |||
BC129087.1 | - | 1291 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
