Assay Details
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080896.1 | NP_543172.1 | 6 - 7 | 753 | 103 | |
GenBank mRNA | AF367468.1 | - | 6 - 7 | 753 | 103 | |
BC099792.1 | - | 832 | 103 | |||
BC129087.1 | - | 971 | 103 | |||
FQ212957.1 | - | 3 - 4 | 332 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
