Assay Details
Gene Symbol: Fgf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130814.1 | NP_570827.1 | 4 - 5 | 607 | 92 | |
GenBank mRNA | AB079374.1 | - | 4 - 5 | 607 | 92 | |
AF348446.1 | - | 3 - 4 | 421 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
