Assay Details
Gene Symbol: Slc25a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133614.2 | NP_598298.1 | 5 - 6 | 582 | 89 | |
XM_006240114.2 | XP_006240176.1 | 5 - 6 | 1908 | 89 | ||
GenBank mRNA | AJ289714.1 | - | 5 - 6 | 426 | 89 | |
BC089099.1 | - | 5 - 6 | 582 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
