Assay Details
Gene Symbol: Podxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138848.1 | NP_620203.1 | 5 - 6 | 1178 | 64 | |
XM_008762758.1 | XP_008760980.1 | 5 - 6 | 1338 | 64 | ||
GenBank mRNA | AB020726.1 | - | 5 - 6 | 1251 | 64 | |
AF109393.1 | - | 5 - 6 | 1178 | 64 | ||
BC070886.1 | - | 5 - 6 | 1248 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
