Assay Details
Gene Symbol: Slc36a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139339.1 | NP_647555.1 | 3 - 4 | 450 | 62 | |
GenBank mRNA | AF512430.1 | - | 3 - 4 | 450 | 62 | |
BC166414.1 | - | 3 - 4 | 384 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
