Assay Details
Gene Symbol: Slc36a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139339.1 | NP_647555.1 | 4 - 5 | 535 | 95 | |
GenBank mRNA | AF512430.1 | - | 4 - 5 | 535 | 95 | |
BC166414.1 | - | 4 - 5 | 469 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
