Assay Details
Gene Symbol: Gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183326.2 | NP_899155.1 | 1 - 2 | 213 | 99 | |
GenBank mRNA | BC100061.1 | - | 1 - 2 | 213 | 99 | |
FQ211937.1 | - | 1 - 2 | 466 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC100061.1, NM_183326.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
