Assay Details
Gene Symbol: Hyal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207616.1 | NP_997499.1 | 2 - 3 | 947 | 91 | |
GenBank mRNA | AB100600.1 | - | 2 - 3 | 947 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
