Assay Details
Gene Symbol: Cdc42se1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039044.1 | NP_001034133.1 | 4 - 5 | 536 | 99 | |
GenBank mRNA | BC091418.1 | - | 4 - 5 | 536 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
