Assay Details
Gene Symbol: Rps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009239.1 | NP_001009239.1 | 4 - 5 | 369 | 86 | |
GenBank mRNA | BC088450.1 | - | 4 - 5 | 369 | 86 | |
X51536.1 | - | 4 - 5 | 375 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
