Assay Details
Gene Symbol: Ahsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107241.1 | NP_001100711.1 | 94 | |||
XM_006251539.2 | XP_006251601.1 | 94 | ||||
XM_006251540.2 | XP_006251602.1 | 94 | ||||
XM_006251541.2 | XP_006251603.1 | 94 | ||||
XM_006251542.2 | XP_006251604.1 | 94 | ||||
GenBank mRNA | BC099114.1 | - | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107241.1 transcripts.
