Assay Details
Gene Symbol: Fut10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012360.1 | NP_001012360.1 | 3 | 1220 | 79 | |
GenBank mRNA | AJ879584.1 | - | 3 | 1220 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
