Assay Details
Gene Symbol: Cd244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250259.2 | XP_006250321.1 | 61 | |||
GenBank mRNA | AF209406.1 | - | 61 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250259.2 | XP_006250321.1 | 61 | |||
GenBank mRNA | AF209406.1 | - | 61 |