Assay Details
Gene Symbol: Cdk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053593.2 | NP_446045.1 | 2 - 3 | 404 | 62 | |
GenBank mRNA | BC070956.1 | - | 2 - 3 | 404 | 62 | |
FQ214013.1 | - | 2 - 3 | 420 | 62 | ||
FQ219275.1 | - | 2 - 3 | 380 | 62 | ||
FQ234663.1 | - | 1 - 2 | 33 | 62 | ||
L11007.1 | - | 2 - 3 | 336 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
