Assay Details
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017061.2 | NP_058757.1 | 4 - 5 | 1042 | 81 | |
XM_006254713.2 | XP_006254775.1 | 2 - 3 | 1125 | 81 | ||
XM_006254714.2 | XP_006254776.1 | 3 - 4 | 1025 | 81 | ||
XM_006254715.2 | XP_006254777.1 | 2 - 3 | 1126 | 81 | ||
GenBank mRNA | BC078861.1 | - | 4 - 5 | 1042 | 81 | |
FQ230374.1 | - | 2 - 3 | 277 | 81 | ||
FQ231737.1 | - | 1 - 2 | 66 | 81 | ||
U11038.1 | - | 2 - 3 | 1010 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
