Assay Details
Gene Symbol: Il1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017019.1 | NP_058715.1 | 6 | 677 | 101 | |
GenBank mRNA | AJ245642.1 | - | 6 | 641 | 101 | |
AJ245643.1 | - | 4 | 442 | 101 | ||
D00403.1 | - | 6 | 677 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
