Assay Details
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013110.2 | NP_037242.2 | 2 - 3 | 157 | 82 | |
XM_006232139.2 | XP_006232201.1 | 2 - 3 | 3003 | 82 | ||
XM_008760860.1 | XP_008759082.1 | 2 - 3 | 3003 | 82 | ||
GenBank mRNA | AF010464.1 | - | 2 - 3 | 166 | 82 | |
AF367210.1 | - | 2 - 3 | 157 | 82 | ||
FQ226549.1 | - | 727 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
