Assay Details
Gene Symbol: FOXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021084213.1 | XP_020939872.1 | 2433 | 94 | ||
XM_021084221.1 | XP_020939880.1 | 2324 | 94 | |||
XM_021084231.1 | XP_020939890.1 | 2285 | 94 | |||
GenBank mRNA | AK395232.1 | - | 1 - 2 | 650 | 94 | |
DQ243691.2 | - | 1 - 2 | 182 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
